Masterflex® Single-Use Replacement Pump Heads for Quattroflow Pump Systems
Disposable Pump Chamber, PP machined for ABS and stainless steel single-use Quattroflow pump systems
Catalog # MFLX76000-00
Supplier: Avantor Fluid Handling
Masterflex® Single-Use Replacement Pump Heads for Quattroflow Pump Systems
Catalog # MFLX76000-00
Supplier: Avantor Fluid Handling
Jump to:
Product Details & Documents
Keep your Quattroflow™ pump system up and running with these quality replacement pump heads.
- No mechanical seal or rotating parts that can fail—only the diaphragm and body come in contact with the fluid
- Single-use pump heads are constructed with a disposable polypropylene chamber
- Reusable pump heads are constructed of durable 316L stainless steel
Keep your Quattroflow pump system up and running with these quality replacement pump heads. The quaternary diaphragm design enables risk-free dry-running, self-priming, and a high turn-down ratio. These pump heads can safely and reliably pump plasma products, proteins, monoclonal antibodies, vaccines, and more.
Order the Masterflex® Single-use Quattroflow™ Pump Systems separately.
Specifications
- Description:Disposable Pump Chamber, PP machined for ABS and stainless steel single-use Quattroflow pump systems
- Seq:01
- Cat. No.:MFLX76000-00
- Warranty:Shelf life
- Inlet connection:1⁄4" Tri-Clamp
- Wetted materials:PP (housing, valve plate), TPE (diaphragm), EPDM (valves, O-rings)
- Outlet connection:1⁄4" Tri-Clamp
Accessories
Product Information
- Catalog #Electrical plug typeMaximum Flow RateAvailabilityPrice
- Multi-use Quattroflow™ Pump System, 3 L/min, Stainless Steel, with EtherNet/IP™; 115/230 VACMulti-use Quattroflow™ Pump System, 3 L/min, Stainless Steel, with EtherNet/IP™; 115/230 VACMFLX74000-49No plug3 L/minLog in for availability1 * 1 ST - 13 250,00 €



