ContactUsLinkComponent
Masterflex® Fitting, Polypropylene, Elbow, Hosebarb to Thread, 5/16" ID x 1/8" NPT(M); 100/PK
Masterflex® Fitting, Polypropylene, Elbow, Hosebarb to Thread, 5/16" ID x 1/8" NPT(M); 100/PK
Catalog # MFLX40626-86
CE certificate
undefined
Masterflex® Fitting, Polypropylene, Elbow, Hosebarb to Thread, 5/16" ID x 1/8" NPT(M); 100/PK
Catalog # MFLX40626-86
CE certificate
About this item
Quick and easy assembly.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Fitting, Polypropylene, Tee, Hose Barb to Threaded Adapter, 3/4" ID x 3/4" ID x 3/4" NPT(M); 100/PK
Barbed elbow fittings are designed for quick and easy assembly without tools. Combine these simple and economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30 psi).
  • Barb size/tubing int.Ø (port 2):
    5/16"
  • Connection Type:
    Thread to Barb
  • Flow pattern:
    Elbow
  • Thread size NPT(M) (port 1):
    1/8"
  • Description:
    Fitting, Polypropylene, Elbow, Hosebarb to Thread, 5/16" ID x 1/8" NPT(M); 100/PK
  • Seq:
    22
  • Environmentally Preferable:
  • Model:
    40626-86
  • Orifice dimensions:
    0,1875"
  • Sterilisation:
    Autoclave or ethylene oxide (ETO)
  • Pk:
    100
  • Fitting material:
    Polypropylene
Frequently Bought Together