ContactUsLinkComponent
Masterflex® Fitting, Natural Polypropylene, Tee, Hose Barb to Threaded Adapter, 3/4" ID x 3/4" ID x 1/2" NPT(M); 10/PK
Masterflex® Fitting, Natural Polypropylene, Tee, Hose Barb to Threaded Adapter, 3/4" ID x 3/4" ID x 1/2" NPT(M); 10/PK
Catalog # MFLX40610-83
CE certificate
undefined
Masterflex® Fitting, Natural Polypropylene, Tee, Hose Barb to Threaded Adapter, 3/4" ID x 3/4" ID x 1/2" NPT(M); 10/PK
Catalog # MFLX40610-83
CE certificate
About this item
Quick and easy assembly.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
These tee adapters help you install your flexible tubing lines into more rigid, threaded lines. Choose the size and material to suit your application needs.
  • Barb size/tubing int.Ø (port 2):
    ¾"
  • Connection Type:
    Thread to Barb
  • Flow pattern:
    T
  • Description:
    Fitting, Natural Polypropylene, Tee, Hose Barb to Threaded Adapter, ¾" ID × ¾" ID × ½" NPT(M); 10/PK
  • Seq:
    10
  • Environmentally Preferable:
  • Model:
    40610-83
  • Orifice dimensions:
    0,5625"
  • Sterilisation:
    Ethylene oxide (ETO) or gamma
  • Warranty:
    Out of box
  • Pk:
    10
  • Fitting material:
    Polypropylene
Frequently Bought Together