Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
REAGENT RESERVOIRS25ML 1 * 50 items
Catalog # 613-7454
Supplier:
THERMO MATRIX TECHNOLOGIES
Expand image
REAGENT RESERVOIRS25ML 1 * 50 items
Catalog # 613-7454
Supplier:
THERMO MATRIX TECHNOLOGIES
Related Information
Documentation
CoC_MATR8096-11_21181672
Documentation
Frequently Bought Together
VWR® Reagent Reservoirs
Supplier: VWR Collection
Single-use syringes, 3-piece, HENKE-JECT®
Supplier: HENKE SASS WOLF
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Ethanol, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals