HexArmor® Safety Gloves, Helix® 2076, 60660
Catalog # UVEX6066005
Supplier: UVEX
Specifications
- Glove size:5
- Ambidextrous / hand-specific:Hand-specific
- Disposable / reusable:Disposable
- Powdered / powder-free:Powder-free
- Colour:Blue/black
- Seq:1
- EN norm:EN 388:2016
Specifications
About this item
Helix® 2076 gloves feature a 13-gauge HPPE, steel, and fibreglass blend shell with seamless construction that gives wearers' hands advanced cut protection, comfort, and breathability on the job, plus a reinforced thumb saddle patch and a flexible PU palm coating.
- HPPE and steel fiberglass blend shell offering 360° cut protection
- Flexible polyurethane palm coating
- Reinforced thumb crotch area for increased durability
- Seamless construction delivers high comfort and dexterity
The 13-gauge HPPE and steel blend shell offers 360° cut protection and excellent sensitivity in cut protection level F. Flexible polyurethane palm coating provides superior grip and abrasion resistance. A knit wrist helps prevent dirt and debris from entering the glove. Seamless construction allows enhanced comfort while maintaining dexterity and tactility.
Frequently Bought Together