(1R,2S,6R)-2-{[2-(3-FLUOROPHENYL)ETHYL]A 1 * 5 mg
Catalog # MSPPLAS57261761-5
Supplier: MARKET SOURCE PART PROCESS
(1R,2S,6R)-2-{[2-(3-FLUOROPHENYL)ETHYL]A 1 * 5 mg
Catalog # MSPPLAS57261761-5
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together
n-Pentane ≥99.8%, SupraSolv® for GC-ECD/FID, Supelco®
Supplier: Merck
Ethylene glycol ≥98%, TECHNICAL
Supplier: VWR Chemicals
Dichloromethane ≥99.8%, SupraSolv® for GC-MS
Supplier: Merck
Acetone, EMPARTA® ACS analytical reagent, Supelco®
Supplier: Merck
2-Propanol ≥98%, TECHNICAL
Supplier: VWR Chemicals
Ethanol eurodenatured ≥96%, GPR RECTAPUR® (1:1:1)
Supplier: VWR Chemicals
N-Ethyldiisopropylamine ≥99.5% for peptide synthesis
Supplier: VWR Chemicals
Acetonitrile, ChromAR® for HPLC, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Cyclohexane ≥99.8%, SupraSolv® for GC-MS
Supplier: Merck
Methanol ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Dichloromethane ≥99.5% stabilised, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Dichloromethane, anhydrous (max. 0.001% H₂O) ≥99.8% stabilised
Supplier: VWR Chemicals
Methanol, anhydrous (max. 0.002% H₂O) ≥99.9%
Supplier: VWR Chemicals
n-Hexane ≥99%, Reagent Grade
Supplier: Honeywell Chemicals
Chloroform ≥99.8% stabilised ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals