ContactUsLinkComponent
Microscope cover glasses, 24x100 mm, borosilicate glass D 263 M, thickness No. 1.5 (0.16-0.19 mm), 100 pcs./cardboard box, MARIENFELD SUPERIOR 1 * 10.000 items
Catalog # MARI115959
Supplier:  MARIENFELD
Microscope cover glasses, 24x100 mm, borosilicate glass D 263 M, thickness No. 1.5 (0.16-0.19 mm), 100 pcs./cardboard box, MARIENFELD SUPERIOR 1 * 10.000 items
Catalog # MARI115959
Supplier:  MARIENFELD
Frequently Bought Together