ContactUsLinkComponent
CD3 naive helper T cells were isolated from a leukopak by negative selection. After isolation, naive CD3 T cells were tested for pur ity and viability by flow cytometry and then quickly cryopreserved on site in CryoStor® CS10 freeze media (Catalog #: 1 * 10 Mou
Catalog # PB0345RANC-1
CD3 naive helper T cells were isolated from a leukopak by negative selection. After isolation, naive CD3 T cells were tested for pur ity and viability by flow cytometry and then quickly cryopreserved on site in CryoStor® CS10 freeze media (Catalog #: 1 * 10 Mou
Catalog # PB0345RANC-1
Frequently Bought Together