MEDIUM-CHAIN TRIGLYCERIDES NF 1 * 4 L
SPCMM4200-4LT
Frequently Bought Together
n-Hexane ≥99.0%, PESTINORM® for capillary GC analysis
: VWR Chemicals
Water, TECHNICAL, demineralised
: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
1-Butanol ≥98.5%, GPR RECTAPUR®
: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
: VWR Chemicals
Ethanol 96% (v/v), EMPROVE® EXPERT Ph. Eur., JP, USP, SAFC®
: MERCK PRODUCTION CHEMICALS
Diethyl ether ≥99% stabilised, GPR RECTAPUR®
: VWR Chemicals
Toluene ≥99.9% (by GC), SPECTRONORM® for spectroscopy
: VWR Chemicals
Methanol ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
Methanol, anhydrous (max. 0.002% H₂O) ≥99.9%
: VWR Chemicals
2-Propanol ≥99.9%, HiPerSolv CHROMANORM® for LC-MS
: VWR Chemicals