Recombinant sel-tagged synthethic selenoproteins containing Sec. Enables purification of PAO-Sepharose and Sec-specific modification s using covalent targeting with electrophilic moieties, eg. for fluorescence labeling or radiolabeling. More than 90% 1 * 1 AU
Catalog # SELZSEL-TAGCUSTOM
Supplier: SELENOZYME
Frequently Bought Together