ContactUsLinkComponent
Seat stand up Polyamide, 5-branch, reinforced base with heavy-duty nylon casters Black gas-lift column with height adjustment from 470 to 600 mm and shock absorber Ergonomic seat in black polyurethane, tilted forward, with carry handle 1 * 1 items
Catalog # BIEN7NA20NBNR00905
Supplier:  Kango
Seat stand up Polyamide, 5-branch, reinforced base with heavy-duty nylon casters Black gas-lift column with height adjustment from 470 to 600 mm and shock absorber Ergonomic seat in black polyurethane, tilted forward, with carry handle 1 * 1 items
Catalog # BIEN7NA20NBNR00905
Supplier:  Kango
Frequently Bought Together