ContactUsLinkComponent
DH.WEB01004 Multi-purpose (Extraction) Water Bath WEB-4, with 4 places ? 110 mm, temperature range: ambient +5C to 100C, timer: 99 hr. / 59 min., bath dim.: 610 x 160 x 160 mm, heater: 1,4 kW, power supply: 230 V, 50/60 Hz 1 * 1 items
Catalog # WITGDH.WEB01004
Supplier:  witeg Labortechnik
DH.WEB01004 Multi-purpose (Extraction) Water Bath WEB-4, with 4 places ? 110 mm, temperature range: ambient +5C to 100C, timer: 99 hr. / 59 min., bath dim.: 610 x 160 x 160 mm, heater: 1,4 kW, power supply: 230 V, 50/60 Hz 1 * 1 items
Catalog # WITGDH.WEB01004
Supplier:  witeg Labortechnik
Frequently Bought Together