Order Entry
Belgium
Orders LinkContactUsLinkComponent
Phenol

Phenol

500 mL
Catalog # ACSDF330654.L5
Supplier:   CPACHEM
Phenol
Catalog # ACSDF330654.L5
Supplier:  CPACHEM
CAS Number:  108-95-2
Phenol

500 mL

CAS Number:  108-95-2
Price
356,00 €
Log in for availability
1 * 500 mL
Product Details & Documents
Synonyms:  Phenylalcohol, Carbolic acid, Hydroxybenzene
  • CAS Number:108-95-2
  • Formula:C₆H₅OH
  • MDL Number:MFCD00002143
  • Molecular Weight:94.11 g/mol
  • Density:1 g/cm³
Specifications

Specifications

  • Pk:
    500 mL

Specifications