Order Entry
Belgium
Orders LinkContactUsLinkComponent
Dropping pipettes, disposable

Dropping pipettes, disposable

Disposable pipette, 1,8 ml, 98 mm
Catalog # BURK7555-0001
Supplier:   BURKLE
Dropping pipettes, disposable
Catalog # BURK7555-0001
Supplier:  BURKLE
Dropping pipettes, disposable
Dropping pipettes, disposable

Disposable pipette, 1,8 ml, 98 mm

Price
38,90 €
Log in for availability
1 * 100 ST
Product Details & Documents

Made of LDPE.

  • Ungraduated
Specifications
  • Capacity:
    1,8 ml
  • Description:
    Disposable pipette
  • Length:
    98 mm
  • Seq:
    01
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.