Clog, shoe and boot rack storage
Boot rack, wall-mounted, for 16 pairs, 1184×332×1605 mm
Catalog # TEKMCRFS-BRWM16
Supplier: TEKNOMEK
Jump to:
Product Details & Documents
Our hygienically designed stainless steel shoe and boot racks come with a hygienic, easier to clean, continuous piece of prongs.
- Supplied fully welded, ready to install
- 304 grade stainless steel, brushed dull polish finish
- Free standing with two stainless steel levelling feet and non-marking rubber base and front
- Wall mounted option to save space
- Two fully welded stainless steel plates at the rear
- Mobile version also available!
With a narrower and taller design, it provides enough space so they can be thoroughly cleaned on the rack. Choose from boot rack or shoe racks, whether wall mounted to save space, freestanding or mobile for even easier cleaning. Sizes range from 8 to 36 pairs.
Specifications
- Description:Boot rack, wall-mounted, for 16 pairs
- Seq:08
- W×D×H:1184×332×1605 mm