Order Entry
Belgium
Orders LinkContactUsLinkComponent
Dollies

Dollies

Circular dollie, 460 mm Ø stainless steel, 460×460×165 mm
Catalog # TEKMBS0010/460
Supplier:   TEKNOMEK
Dollies
Catalog # TEKMBS0010/460
Supplier:  TEKNOMEK
Dollies
Dollies

Circular dollie, 460 mm Ø stainless steel, 460×460×165 mm

Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents

Our stainless steel circular dollies are a extremely robust, with an up to 150 kg payload.

  • Fully welded 304 grade stainless steel
  • Brushed dull polish finish
  • 4 swivel castors
  • Crevice free
  • Sturdy design
  • Easy-clean design
  • Fully welded, ready to use

They come with swivel castors, ensuring smooth and effortless movement. Rectangular stainless steel and aluminum bogies are also available.

Specifications
  • Description:
    Circular dollie, 460 mm Ø stainless steel
  • Seq:
    04
  • W×D×H:
    460×460×165 mm
Recommandations

Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".

Dans le cas contraire, vous recevrez des recommandations génériques.