Store below 30C in tightly closed container and prepared medium at 2-8C 1 * 500 g
HIMEM1852I-500G
: HIMEDIA
Store below 30C in tightly closed container and prepared medium at 2-8C 1 * 500 g
HIMEM1852I-500G
: HIMEDIA
Frequently Bought Together
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection
VWR® Facial Tissues
: VWR Collection
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection