Order Entry
Belgium
Orders LinkContactUsLinkComponent
Cryogenic warning labels

Cryogenic warning labels

Biohazard labels, Plastic, Biohazard Picto, 50,8×50,8 mm, 100 labels/roll
Catalog # GAINWLA-006-0.1P
Supplier:   GA
Cryogenic warning labels
Catalog # GAINWLA-006-0.1P
Supplier:  GA
Cryogenic warning labels
Cryogenic warning labels

Biohazard labels, Plastic, Biohazard Picto, 50,8×50,8 mm, 100 labels/roll

Price
45,60 €
Log in for availability
1 * 1 ROU
Product Details & Documents

Cryogenic warning labels for storage in liquid nitrogen, freezers, dry ice and other harsh environments. It is resistant to multiple freeze-thaw cycles, boiling water, UV rays, sterilization by gamma irradiation, deep-freeze and harsh lab environments.

  • Water resistant
  • Wear and tear-proof
  • Laminated
  • No perforation

The pictograms in laminated labels are protected against water, alcohol, chemicals and solvents. Temperature range from −196 to +100 °C.

Specifications
  • Description:
    Biohazard labels
  • Dimensions:
    50,8×50,8 mm
  • Material:
    Plastic
  • Seq:
    12
  • Packed:
    100 labels/roll
  • Legend:
    Biohazard Picto