Cryogenic warning labels
Removable warning symbol labels, black, Plastic, Radioactive Picto, 12,7 mm, 1000 labels/roll
GAINWL-013-1P
: GA
Cryogenic warning labels for storage in liquid nitrogen, freezers, dry ice and other harsh environments. It is resistant to multiple freeze-thaw cycles, boiling water, UV rays, sterilization by gamma irradiation, deep-freeze and harsh lab environments.
- Water resistant
- Wear and tear-proof
- Laminated
- No perforation
The pictograms in laminated labels are protected against water, alcohol, chemicals and solvents. Temperature range from −196 to +100 °C.
- Description:Removable warning symbol labels, black
- Dimensions:12,7 mm
- Material:Plastic
- Seq:02
- Packed:1000 labels/roll
- Legend:Radioactive Picto
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.



