Order Entry
Belgium
Orders LinkContactUsLinkComponent
ISO warning labels

ISO warning labels

ISO warning label - Low Temperature, Black/yellow, 19,05×19,05 mm, 250 labels/roll
Catalog # GAINWL-010-0.25R
Supplier:   GA
ISO warning labels
Catalog # GAINWL-010-0.25R
Supplier:  GA
ISO warning labels
ISO warning labels

ISO warning label - Low Temperature, Black/yellow, 19,05×19,05 mm, 250 labels/roll

Price
49,80 €
Log in for availability
1 * 1 ROU
Product Details & Documents

Pre-printed general warning labels for the identification of equipment, containers, and material that may pose a safety risk in the laboratory environment.

  • Withstand high-temperature conditions up to 100° C (212° F)
  • Pre-printed with full color, yellow on silver background, design
  • Durable thermoplastic material is resistant to wear and tear, water, mild chemicals, and detergents
  • Resistant to gamma irradiation (tested to up to 50kGy)

Coated with a glove-friendly adhesive, these waterproof thermoplastic labels are perfect for labeling water baths, heat blocks, autoclave containers, and instruments or tools with moving parts.

Specifications
  • Colour:
    Black/yellow
  • Description:
    ISO warning label - Low Temperature
  • Dimensions:
    19,05×19,05 mm
  • Seq:
    02
  • Packed:
    250 labels/roll
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.