Cryogenic warning labels
Removable warning symbol labels, Plastic, Hot Surface Hazard Picto, 19,05×19,05 mm, 250 labels/roll
GAINWL-008-1R
: GA
Cryogenic warning labels for storage in liquid nitrogen, freezers, dry ice and other harsh environments. It is resistant to multiple freeze-thaw cycles, boiling water, UV rays, sterilization by gamma irradiation, deep-freeze and harsh lab environments.
- Water resistant
- Wear and tear-proof
- Laminated
- No perforation
The pictograms in laminated labels are protected against water, alcohol, chemicals and solvents. Temperature range from −196 to +100 °C.
- Description:Removable warning symbol labels
- Dimensions:19,05×19,05 mm
- Material:Plastic
- Seq:04
- Packed:250 labels/roll
- Legend:Hot Surface Hazard Picto



