Order Entry
Belgium
Orders LinkContactUsLinkComponent
Cryogenic warning labels

Cryogenic warning labels

Non-hazardous labels, Plastic, —, 44,45×19,05 mm, 250 labels/roll
Catalogus # GAINWL-002-0.25P
Leverancier:   GA
Cryogenic warning labels
Catalogus # GAINWL-002-0.25P
Leverancier:  GA
Cryogenic warning labels
Cryogenic warning labels

Non-hazardous labels, Plastic, —, 44,45×19,05 mm, 250 labels/roll

Price
34,20 €
Log in voor beschikbaarheid
1 * 1 ROU
Product Details & Documents

Cryogenic warning labels for storage in liquid nitrogen, freezers, dry ice and other harsh environments. It is resistant to multiple freeze-thaw cycles, boiling water, UV rays, sterilization by gamma irradiation, deep-freeze and harsh lab environments.

  • Water resistant
  • Wear and tear-proof
  • Laminated
  • No perforation

The pictograms in laminated labels are protected against water, alcohol, chemicals and solvents. Temperature range from −196 to +100 °C.

Specificaties
  • Description:
    Non-hazardous labels
  • Dimensions:
    44,45×19,05 mm
  • Material:
    Plastic
  • Seq:
    10
  • Packed:
    250 labels/roll
  • Legend: