Orders LinkContactUsLinkComponent
Laboratory warning labels
Laboratory warning labels
Laboratory warning labels, 1×1,5"
Catalog # GAINL-001-1R
Supplier:   GA
Laboratory warning labels
Catalog # GAINL-001-1R
Supplier:  GA
Laboratory warning labels
Laboratory warning labels
Laboratory warning labels, 1×1,5"
Price
31,40 €
Log in for availability
1 * 1 ROU
Product Details & Documents
Preprinted, and blank warning labels for the identification of equipment, containers, and material.
  • Coated with either a permanent or removable adhesive
  • Waterproof, low and high-temperature resistant
  • Can be applied on plastic, glass, cardboard, and metal surfaces
Labels are designed to alert personnel to safety risks in the laboratory environment, and to communicate special use, handling, and disposal. With a wide temperature resistance range, these labels can be used in lab freezers and refrigerators, at room temperature (doors, drawers, walls, instruments, and so on), hot surfaces, including water baths and heating blocks, as well as on equipment, instruments, and tools with moving parts in order to minimise injury risks.
Specifications
  • Description:
    Laboratory warning labels
  • Dimensions:
    1×1,5"
  • Seq:
    01
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.