Tork Heavy-Duty Cloth Folded, nonwoven, white, widthe 35.5cm, sheet-length 41.5cm, 105 sheets, system W4 1 * 4 items
Catalog # SCAA530179_U
Supplier: Essity
Tork Heavy-Duty Cloth Folded, nonwoven, white, widthe 35.5cm, sheet-length 41.5cm, 105 sheets, system W4 1 * 4 items
Catalog # SCAA530179_U
Supplier: Essity
Frequently Bought Together
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Ethanol absolute ≥99.5% USP, Ph. Eur.
Supplier: VWR Chemicals
Ethanol 96% vol Ph. Eur.
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Pipette tips, Finntip™
Supplier: Thermo Fisher Scientific
Trifluoroacetic acid, Uvasol® for spectroscopy, Supelco®
Supplier: Merck
VWR® Facial Tissues
Supplier: VWR Collection
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Single-use syringes, 3-piece, HENKE-JECT®
Supplier: HENKE SASS WOLF