Order Entry
Belgium
Orders LinkContactUsLinkComponent
Stirring rods

Stirring rods

Stirring rods, glass, with fused ends, Glass, 5 mm, 150 mm
Catalog # HECH42850012
Stirring rods
Catalog # HECH42850012
Stirring rods
Stirring rods

Stirring rods, glass, with fused ends, Glass, 5 mm, 150 mm

Price
54,40 €
Log in for availability
1 * 100 ST
Product Details & Documents

AR-Glas®. With fused ends.

Specifications
  • Ø:
    5 mm
  • Description:
    Stirring rods, glass, with fused ends
  • Length:
    150 mm
  • Material:
    Glass
  • Seq:
    012