ContactUsLinkComponent
Precision ASTM hydrometer type 314H-82, ASTM E100-19, density range: 750-800 in 0.5 kg/m , total length: 330 mm, body diameter 24 mm, without thermometer, adjusting temperature: 15C, including works-calibration certificate with 3 calibration points 1 * 1 items
Catalog # SCNL3222154/03/3P
Precision ASTM hydrometer type 314H-82, ASTM E100-19, density range: 750-800 in 0.5 kg/m , total length: 330 mm, body diameter 24 mm, without thermometer, adjusting temperature: 15C, including works-calibration certificate with 3 calibration points 1 * 1 items
Catalog # SCNL3222154/03/3P
Frequently Bought Together