0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Belgium
ContactUsLinkComponent
KIT, MEDIUM FORMULATED AT NORMAL BLOOD GLUCOSE LEVEL WITHOUT SERUM AND WITH ANIMAL GROWTH FACTOR CULTUREBOOST 1 * 1 KIT
CELS4N3-500
:
CELL SYSTEMS
KIT, MEDIUM FORMULATED AT NORMAL BLOOD GLUCOSE LEVEL WITHOUT SERUM AND WITH ANIMAL GROWTH FACTOR CULTUREBOOST 1 * 1 KIT
CELS4N3-500
:
CELL SYSTEMS
Frequently Bought Together
Laboratory trays
: KARTELL
Serological pipettes, Stripette™
: Corning
Eppendorf® DNA LoBind® Microcentrifuge Tubes
: EPPENDORF
VWR®, Ice Pans, with or without Lid
: VWR Collection
Eppendorf Tubes® Microtubes with Snap Cap, 5,0 ml
: EPPENDORF
VWR®, Petri Dish, PS
: VWR Collection
VWR® Water Sampling Bottles, with Tamper Evident Cap
: VWR Collection
Self-standing centrifuge tubes, 50 ml, Corning®
: Corning
VWR®, Transfer Pipettes, High Performance
: VWR Collection
Centrifuge tubes, Falcon®
: Corning
VWR®, Cryoboxes, Cardboard, 133×133 mm
: VWR Collection
VWR®, Cryoboxes with Dividers
: VWR Collection
VWR® Serological Pipette, Standard Line
: VWR Collection
Cryoboxes, 133×133 mm
: TENAK
Cell culture flasks, Nunc™
: Thermo Fisher Scientific
VWR® Standard Line Transfer Pipettes
: VWR Collection