ContactUsLinkComponent
COLUMN COGENT C30 5UM 4.6 X 150MM 200A 1 * 1 items
Catalogus # MICS72530-15P
Leverancier:  MICROSOLV
COLUMN COGENT C30 5UM 4.6 X 150MM 200A 1 * 1 items
Catalogus # MICS72530-15P
Leverancier:  MICROSOLV
Frequently Bought Together