Orders LinkContactUsLinkComponent
Quick-lock connectors
Quick-lock connectors
Quick-lock connector (m), includes 3× PTFE-ferrule (1,6/2,3/3,2 mm ID), 1 Safety Cap
Catalog # SCTV360180
Supplier:   S.C.A.T.
Quick-lock connectors
Catalog # SCTV360180
Supplier:  S.C.A.T.
Quick-lock connectors
Quick-lock connectors
Quick-lock connector (m), includes 3× PTFE-ferrule (1,6/2,3/3,2 mm ID), 1 Safety Cap
Price
37,60 €
Log in for availability
1 * 1 ST
Product Details & Documents
Easy and safe connection of capillary tubing and SCAT Safety Caps using quick couplings. Connect and disconnect with a "click" - without dripping or air bubbles. The valve closes automatically when disconnected. Thanks to the included PFA fittings, all common hose diameters can be connected.
  • Made of chemically resistant glass-filled polypropylene, PEEK® materials
  • Leak protection thanks to integrated shut-off valves
  • Closed system for drip-free disconnection with very low air entrapment
  • Minimizes contamination and no need to purge air bubbles from the system
  • Suitable for capillaries with outer diameter 1,6/2,3/3,2 mm
  • User-friendly, ergonomic design, enables safe connection
  • Up to vacuum 15 psi, 1,0 bar pressure resistance
The quick-lock connector sets are suitable for connecting 2 capillaries (Set 1) or one capillary and one SCAT Safety Cap (Set 2).
Specifications
  • For:
    1 Safety Cap
  • Description:
    Quick-lock connector (m), includes 3× PTFE-ferrule (1,6/2,3/3,2 mm ID)