Corning® 25-Layer CellCube® Module, Corning
Product Details & Documents
The CellCube Module is an integral, encapsulated, sterile, single-use device that is 100% pressure-hold tested before shipment.
It is comprised of a series of parallel, styrene plates joined to create thin, sealed, laminar flow spaces between adjacent plates. Each culture plate receives proprietary tissue culture treatment prior to assembly to ensure dense, uniform cell growth. These CellCube Modules provide: a series of parallel, polystyrene plates joined to create thin, sealed, laminar flow spaces between adjacent plates, 21,250 cm² polystyrene growth surface optimally treated for the growth of attachment dependent cells, a perfused environment which more closely simulates in vivo conditions and reliably distributes nutrients and oxygen, an integral, encapsulated, sterile, single-use device that is 100% pressure-hold tested before shipment. Square Flask Shape.
Documents
Certificates
Search by Lot NumberSpecifications
- Description:25-layer CellCube® module with 21,250 cm² growth surface, Polycarbonate/Polystyrene, Sterile
- Seq:02
- Growth area:21,250 cm²
- Cat. No.:CORN3304
- Media volume:1500 ml
- Surface:Corning® CellBIND®
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.



