ContactUsLinkComponent
Custom Cat. No. SP-2846-GS FluidX 48-format, 1.9ml ExternalThread, Next-Gen Jacket Tri-coded CryoTube, 2D Code on base, 1D LinearBarcode and Human Readable Numberon side, capped, Gamma Sterilised, 8racks per case, Double Bagged in rowsof 4 racks 48-f 1 * 384 items
Catalog # BCISSP-2846-GS
Supplier:  BIOCISION
Custom Cat. No. SP-2846-GS FluidX 48-format, 1.9ml ExternalThread, Next-Gen Jacket Tri-coded CryoTube, 2D Code on base, 1D LinearBarcode and Human Readable Numberon side, capped, Gamma Sterilised, 8racks per case, Double Bagged in rowsof 4 racks 48-f 1 * 384 items
Catalog # BCISSP-2846-GS
Supplier:  BIOCISION
Frequently Bought Together