ContactUsLinkComponent
OPERATOR IDENTIFICATION. ALLOWS INDIVIDUAL OPERATORS TO BE IDENTIFIED ON LOG RECORDS AND PRINTOUTS BY ASSIGNING USER NUMBERS THAT MUST BE ENTERED BEFORE A CYCLE CAN START. 1 * 1 items
Catalog # PRIOPC/OPR/IDN
Supplier:  Priorclave
OPERATOR IDENTIFICATION. ALLOWS INDIVIDUAL OPERATORS TO BE IDENTIFIED ON LOG RECORDS AND PRINTOUTS BY ASSIGNING USER NUMBERS THAT MUST BE ENTERED BEFORE A CYCLE CAN START. 1 * 1 items
Catalog # PRIOPC/OPR/IDN
Supplier:  Priorclave
Frequently Bought Together