Order Entry
Belgium
Orders LinkContactUsLinkComponent
γ-Cyclodextrine 98%

γ-Cyclodextrine 98%

Bottle, 500 g
Catalog # APOSBIB6136-500G
Irritant
Supplier:   Apollo Scientific
γ-Cyclodextrine 98%
Catalog # APOSBIB6136-500G
Supplier:  Apollo Scientific
CAS Number:  17465-86-0
γ-Cyclodextrine 98%

Bottle, 500 g

CAS Number:  17465-86-0
Price
397,00 €
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  gamma-Cyclodextrin, Cyclooctaamylose, γ-Cyclodextrin, Schardinger gamma-Dextrin
  • CAS Number:17465-86-0
  • Formula:C₄₈H₈₀O₄₀
  • MDL Number:MFCD00009595
  • Molecular Weight:1297.1248 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 g

Specifications