Order Entry
Belgium
Orders LinkContactUsLinkComponent
D-(+)-Mannose 99,5%

D-(+)-Mannose 99,5%

Bottle, 500 g
Catalog # APOSBIM0134-500G
Irritant
Supplier:   Apollo Scientific
D-(+)-Mannose 99,5%
Catalog # APOSBIM0134-500G
Supplier:  Apollo Scientific
CAS Number:  3458-28-4
D-(+)-Mannose 99,5%

Bottle, 500 g

CAS Number:  3458-28-4
Price
109,00 €
Log in for availability
1 * 500 g
Product Details & Documents
  • Heavy metals: <10ppm
Synonyms:  (2S,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal
  • CAS Number:3458-28-4
  • Formula:C₆H₁₂O₆
  • MDL Number:MFCD00064122
  • Molecular Weight:180.16 g/mol
  • Melting Pt:133…140 °C
  • Density:1.54 g/cm³ (25 °C)
  • Storage Temperature:Ambient
  • EINECS:222-392-4

Documents

Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.