γ-Cyclodextrine
Bottle, 50 g
Catalog # APOSBIM1016-50G
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Synonyms: gamma-Cyclodextrin, Cyclooctaamylose, γ-Cyclodextrin, Schardinger gamma-Dextrin
- CAS Number:17465-86-0
- Formula:C₄₈H₈₀O₄₀
- MDL Number:MFCD00009595
- Molecular Weight:1297.1248 g/mol
- Storage Temperature:Ambient
Specifications
Specifications
- Pack type:Bottle
- Pk:50 g



