Order Entry
Belgium
Orders LinkContactUsLinkComponent
n-Decyl-β-D-maltoside

n-Decyl-β-D-maltoside

Bottle, 5 g
Catalog # APOSBID6002-5G
Irritant
Supplier:   Apollo Scientific
n-Decyl-β-D-maltoside
Catalog # APOSBID6002-5G
Supplier:  Apollo Scientific
CAS Number:  82494-09-5
n-Decyl-β-D-maltoside

Bottle, 5 g

CAS Number:  82494-09-5
Price
362,00 €
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  DM, N-Decyl-Beta-Maltoside, Decyl 4-O-a-D-glucopyranosyl-b-D-glucopyranoside, n-Decyl-beta-D-maltopyranoside, Decyl b-D-maltopyranoside
  • CAS Number:82494-09-5
  • Formula:C₂₂H₄₂O₁₁
  • MDL Number:MFCD00061624
  • Molecular Weight:482.56 g/mol
  • Storage Temperature:Freezer
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.