Order Entry
Belgium
Orders LinkContactUsLinkComponent
D-(-)-Sorbitol

D-(-)-Sorbitol

Bottle, 1 kg
Catalog # APOSBI2889-1KG
Irritant
Supplier:   Apollo Scientific
D-(-)-Sorbitol
Catalog # APOSBI2889-1KG
Supplier:  Apollo Scientific
CAS Number:  50-70-4
D-(-)-Sorbitol

Bottle, 1 kg

CAS Number:  50-70-4
Price
86,90 €
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  (2R,3R,4R,5S)-Hexane-1,2,3,4,5,6-hexaol, (Sorbitol), D(-)-Glucitol, D-Sorbitol, Sorbitol
  • CAS Number:50-70-4
  • Formula:HOH₂C(CH(OH))₄CH₂OH
  • MDL Number:MFCD00004708
  • Molecular Weight:182.17 g/mol
  • Storage Temperature:Ambient

Documents

Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.