Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

D-(+)-Mannose 99,5%

Bottle, 1 kg
Catalog # APOSBIM0134-1KG
Irritant
Supplier:   Apollo Scientific
D-(+)-Mannose 99,5%
Catalog # APOSBIM0134-1KG
Supplier:  Apollo Scientific
CAS Number:  3458-28-4
D-(+)-Mannose 99,5%

Bottle, 1 kg

CAS Number:  3458-28-4
Price
181,00 €
Log in for availability
1 * 1 kg
Product Details & Documents
  • Heavy metals: <10ppm
Synonyms:  (2S,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal
  • CAS Number:3458-28-4
  • Formula:C₆H₁₂O₆
  • MDL Number:MFCD00064122
  • Molecular Weight:180.16 g/mol
  • Melting Pt:133…140 °C
  • Density:1.54 g/cm³ (25 °C)
  • Storage Temperature:Ambient
  • EINECS:222-392-4

 

Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 kg

Specifications