Order Entry
Belgium
Orders LinkContactUsLinkComponent
DBCO-PEG4-NHS ester

DBCO-PEG4-NHS ester

Bottle
Catalog # APOSBIPG1891-100MG
Irritant
Supplier:   Apollo Scientific
DBCO-PEG4-NHS ester
Catalog # APOSBIPG1891-100MG
Supplier:  Apollo Scientific
CAS Number:  1427004-19-0
DBCO-PEG4-NHS ester

Bottle

CAS Number:  1427004-19-0
Price
1 050,00 €
Log in for availability
1 * 100 mg
Product Details & Documents

Usage: Biochemical/ life science applications.

  • Store at −20 °C
Synonyms:  Dibenzocyclooctyne-PEG4-succinimidyl Ester, 2,5-Dioxo-1-pyrrolidinyl 20-(11,12-didehydrodibenz[b,f]azocin-5(6H)-yl)-17,20-dioxo-4,7,10,13-tetraoxa-16-azaeicosanoate, DBCO-PEG4-succinimidyl Ester, ADIBO-PEG4-NHS, DIBACA-PEG4-NHS
  • CAS Number:1427004-19-0
  • Formula:C₃₄H₃₉N₃O₁₀
  • MDL Number:MFCD26793797
  • Molecular Weight:649,68756 g/mol
  • Storage Temperature:Freezer
Specifications

Specifications

  • Pack type:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.