Valinomycin
Bottle, 50 mg
Catalog # APOSBIV0893-50MG
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Anitbiotic produced by Streptomyces fulvissimus. Valinomycin is highly selective for K+ ions over Na+without influencing the permeability of other ions
- CAS Number:2001-95-8
- Formula:C₅₄H₉₀O₁₈N₆
- MDL Number:MFCD00005114
- Molecular Weight:1111.34 g/mol
- Melting Pt:187…190 °C
- Storage Temperature:Réfrigérateur
- EINECS:217-896-6
- UN:2811
- ADR:6.1,I
Specifications
Specifications
- Type de conditionnement:Bottle
- Cdt:50 mg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



