ContactUsLinkComponent
4 945 201 Centrifuge tubes, nominal volume 10 ml, with round bottom, ungraduated, heavy wall, with screw cap, o.d. 16 mm, length 98 mm 1 * 1 items
Catalog # WITG4.945.201
Supplier:  witeg Labortechnik
4 945 201 Centrifuge tubes, nominal volume 10 ml, with round bottom, ungraduated, heavy wall, with screw cap, o.d. 16 mm, length 98 mm 1 * 1 items
Catalog # WITG4.945.201
Supplier:  witeg Labortechnik
Frequently Bought Together