Order Entry
Belgium
Orders LinkContactUsLinkComponent
Immunoassay Blocking Buffer

Immunoassay Blocking Buffer

Immunoassay blocking buffer, pre-coated, 1000 ml
Catalog # ABCAAB171534-1000
Marketsource
Supplier:   Abcam
Immunoassay Blocking Buffer
Catalog # ABCAAB171534-1000
Supplier:  Abcam
Marketsource
CAS Number:  
Immunoassay Blocking Buffer
Immunoassay Blocking Buffer

Immunoassay blocking buffer, pre-coated, 1000 ml

CAS Number:  
Price
431,00 €
Log in for availability
1 * 1.000 mL
Product Details & Documents

Immunoassay blocking buffer effectively preserves the conformation and activity of dried proteins in immunoassays.

Caution: RUO.

Specifications

Specifications

  • Description:
    Immunoassay blocking buffer, pre-coated
  • Volume:
    1000 ml
  • Cat. No.:
    ABCAAB171534-1000
  • Storage temperature:
    Store at room temperature

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.