Order Entry
Belgium
Orders LinkContactUsLinkComponent
SURE competent cells

SURE competent cells

SURE competent cells, For routine cloning of DNA with secondary structures, ≥5×10⁸ transformants/µg, 5×0,2 ml
Catalog # AGLS200238
Supplier:   AGILENT
SURE competent cells
Catalog # AGLS200238
Supplier:  AGILENT
SURE competent cells

SURE competent cells, For routine cloning of DNA with secondary structures, ≥5×10⁸ transformants/µg, 5×0,2 ml

Price
356,00 €
Log in for availability
1 * 1 mL
Product Details & Documents

SURE 2 competent cells are ideal for routine cloning of DNA with secondary structures.

  • Lack components of the pathways that catalyse the rearrangement and deletion of nonstandard secondary and tertiary structures
  • Allows for blue-white colour screening

Replicating eukaryotic DNA in prokaryotic cells can be problematic. Particular eukaryotic genes may contain inverted repeats or secondary structures, such as Z-DNA, that can be rearranged or deleted by E. coli DNA repair systems.

SURE competent cells are deficient in the E. coli genes involved in the rearrangement and deletion of DNA, thus improving cloning efficiencies of DNA containing irregular structures in prokaryotic cells. SURE cells are restriction minus (McrA-, McrCB-, McrF-, Mrr-, HsdR-) endonuclease (endA) deficient, and recombination (recB recJ) deficient.

Caution: For research use only. Not for use in diagnostic procedures.

Specifications
  • Description:
    SURE competent cells
  • Seq:
    04
  • Pk Info:
    5×0,2 ml
  • Feature:
    For routine cloning of DNA with secondary structures, ≥5×10⁸ transformants/µg
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.