Orders LinkContactUsLinkComponent
SURE competent cells
SURE competent cells
SURE electroporation-competent cells, For cloning DNA with secondary structures, ≥1×10¹⁰ transformants/µg, 5×0,1 ml
Catalog # AGLS200227
Supplier:   AGILENT
SURE competent cells
Catalog # AGLS200227
Supplier:  AGILENT
SURE competent cells
SURE electroporation-competent cells, For cloning DNA with secondary structures, ≥1×10¹⁰ transformants/µg, 5×0,1 ml
Price
355,00 €
Log in for availability
1 * 5 ST
Product Details & Documents
SURE 2 competent cells are ideal for routine cloning of DNA with secondary structures.
  • Lack components of the pathways that catalyse the rearrangement and deletion of nonstandard secondary and tertiary structures
  • Allows for blue-white colour screening
Replicating eukaryotic DNA in prokaryotic cells can be problematic. Particular eukaryotic genes may contain inverted repeats or secondary structures, such as Z-DNA, that can be rearranged or deleted by E. coli DNA repair systems.
SURE competent cells are deficient in the E. coli genes involved in the rearrangement and deletion of DNA, thus improving cloning efficiencies of DNA containing irregular structures in prokaryotic cells. SURE cells are restriction minus (McrA-, McrCB-, McrF-, Mrr-, HsdR-) endonuclease (endA) deficient, and recombination (recB recJ) deficient.
Caution: For research use only. Not for use in diagnostic procedures.
Specifications
  • Description:
    SURE electroporation-competent cells
  • Seq:
    02
  • Pk Info:
    5×0,1 ml
  • Feature:
    For cloning DNA with secondary structures, ≥1×10¹⁰ transformants/µg