ContactUsLinkComponent
Dispenser Point of Care Test Cassettes C-Pette™E Labcon Blood collection tube dispenser for Point of Care testing instruments Dispenses blood drops easy with no need to open collection tubes Dispenses one drop of blood onto test cartridge or ca 1 * 100 items
Catalog # 7625-500-000-9
Supplier:  Labcon
Dispenser Point of Care Test Cassettes C-Pette™E Labcon Blood collection tube dispenser for Point of Care testing instruments Dispenses blood drops easy with no need to open collection tubes Dispenses one drop of blood onto test cartridge or ca 1 * 100 items
Catalog # 7625-500-000-9
Supplier:  Labcon
Frequently Bought Together