Order Entry
Belgium
Orders LinkContactUsLinkComponent
Polypropylene Muff Covers with Elastic

Polypropylene Muff Covers with Elastic

Polypropylene muff cover, 16" (406 mm), fits 8 - 12" (203 - 305 mm) Ø, White
Catalog # 129-3074
Polypropylene Muff Covers with Elastic
Catalog # 129-3074
Polypropylene Muff Covers with Elastic

Polypropylene muff cover, 16" (406 mm), fits 8 - 12" (203 - 305 mm) Ø, White

Price
2 740,00 €
Log in for availability
1 * 500 ST
Product Details & Documents

Keeps cleaned non sterile production hoses protected from external sources of contamination.

  • Manufactured and packed in a controlled cleanroom environment
  • Reduces labour costs
  • Tear and puncture resistant
  • Conveniently packaged in 100 per sealed poly bag

These polypropylene muffs are a great choice in non sterile pharmaceutical manufacturing operations. Can be used as a clean equipment cover to protect process hoses.

Poly bagged in packs of 100 to help ensure cleanliness up until point of use.

Delivery information: Certificates of Conformance sent with each shipment.

Specifications
  • Colour:
    White
  • Description:
    Polypropylene muff cover, 16" (406 mm), fits 8 - 12" (203 - 305 mm) Ø
  • Seq:
    8
Recommandations

Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".

Dans le cas contraire, vous recevrez des recommandations génériques.