Order Entry
Belgium
Orders LinkContactUsLinkComponent
Polypropylene Muff Covers with Elastic

Polypropylene Muff Covers with Elastic

Polypropylene muff cover, 7" (178 mm), fits 2,5 - 4" (63 - 102 mm) Ø, White
Catalog # 129-3070
Polypropylene Muff Covers with Elastic
Catalog # 129-3070
Polypropylene Muff Covers with Elastic

Polypropylene muff cover, 7" (178 mm), fits 2,5 - 4" (63 - 102 mm) Ø, White

Price
2 640,00 €
Log in for availability
1 * 1.000 ST
Product Details & Documents

Keeps cleaned non sterile production hoses protected from external sources of contamination.

  • Manufactured and packed in a controlled cleanroom environment
  • Reduces labour costs
  • Tear and puncture resistant
  • Conveniently packaged in 100 per sealed poly bag

These polypropylene muffs are a great choice in non sterile pharmaceutical manufacturing operations. Can be used as a clean equipment cover to protect process hoses.

Poly bagged in packs of 100 to help ensure cleanliness up until point of use.

Delivery information: Certificates of Conformance sent with each shipment.

Specifications
  • Colour:
    White
  • Description:
    Polypropylene muff cover, 7" (178 mm), fits 2,5 - 4" (63 - 102 mm) Ø
  • Seq:
    4
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.