ContactUsLinkComponent
Holder, set V-8xx, I-100 or pressure gauge on V-300/V-600 Usage: Fixation of vacuum cotroller V-8xx, manometer or interface I-100 to vacuum pump V-300 / V-600. Contents: The rod and spacer plate. 1 * 1 items
Catalog # BUCH11065224
Supplier:  BUCHI
Holder, set V-8xx, I-100 or pressure gauge on V-300/V-600 Usage: Fixation of vacuum cotroller V-8xx, manometer or interface I-100 to vacuum pump V-300 / V-600. Contents: The rod and spacer plate. 1 * 1 items
Catalog # BUCH11065224
Supplier:  BUCHI
Frequently Bought Together