Orders LinkContactUsLinkComponent
Multiports, b.safe
Multiports, b.safe
Tubing connector, 8× capillary connector, 8× fittings for tubing Ext.Ø 1,6 mm, 8× fittings for tubing Ext.Ø 2,2 mm, 8× fittings for tubing Ext.Ø 3,2 mm, NPT ¹/₈"
Catalog # 590-0185
Supplier:   Bohlender
Multiports, b.safe
Catalog # 590-0185
Supplier:  Bohlender
Multiports, b.safe
Multiports, b.safe
Tubing connector, 8× capillary connector, 8× fittings for tubing Ext.Ø 1,6 mm, 8× fittings for tubing Ext.Ø 2,2 mm, 8× fittings for tubing Ext.Ø 3,2 mm, NPT ¹/₈"
Price
169,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Extension of the number of capillary and/or tubing connectors on a b.safe Waste Cap. With PFA Fittings for capillaries with Ext.Ø 1,6/2,2/3,2 mm (thread UNF ¹/₄"), 1 hose connector for tubing Int.Ø 5,5 to 8,0 mm (thread NPT ¹/₈").
  • Excellent chemical resistance
  • Available for a wide range of hose diameters
  • The conical shape gets the connection tight and leak-free
Adaptor to enlarge the number of capillary and tubing connectors of a b.safe waste cap.
Specifications
  • Thread:
    NPT ¹/₈"
  • With:
    8× capillary connector, 8× fittings for tubing Ext.Ø 1,6 mm, 8× fittings for tubing Ext.Ø 2,2 mm, 8× fittings for tubing Ext.Ø 3,2 mm
  • Description:
    Tubing connector
  • Seq:
    08