ContactUsLinkComponent
IBC STATION, POLYETHYLENE, BLUE, WITH GALVANISED GRID, FORKLIFT ACCESSIBLE, SUMP CAPACITY: 1000 LITRES, LOAD CAPACITY IN KG: 2000 KG , DIMENSIONS EXTERIOR: W X D X H (MM) 1370 x 1980 x 690 1 * 1 items
Catalog # MSGA38647
Supplier:  asecos
IBC STATION, POLYETHYLENE, BLUE, WITH GALVANISED GRID, FORKLIFT ACCESSIBLE, SUMP CAPACITY: 1000 LITRES, LOAD CAPACITY IN KG: 2000 KG , DIMENSIONS EXTERIOR: W X D X H (MM) 1370 x 1980 x 690 1 * 1 items
Catalog # MSGA38647
Supplier:  asecos
Frequently Bought Together